Learn More
Cayman Chemical Debio 0123 (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502919251
Encompass_Preferred
A WEE1 inhibitor (IC50 = 0.8 nM); reduces tumor volume in combination etoposide in a PDX mouse model of stomach cancer at 30 mg/kg per day
Catalog No. 50-291-9251 Supplier Cayman Chemical Supplier No. 3967410 MG
A WEE1 inhibitor (IC50 = 0.8 nM); reduces tumor volume in combination etoposide in a PDX mouse model of stomach cancer at 30 mg/kg per day