Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deblock trichloroacetic acid soln., 3% w/w in dichloromethane

Catalog No. AAJ62821K4
Change view
Click to view available options
Quantity:
200 mL
450 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62821K4 2.5 L
AAJ62821AH 200 mL
AAJ62821AO 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62821K4 Supplier Thermo Scientific Chemicals Supplier No. J62821K4
  • $358.50
    $304.65 / Each of 1

    Save $53.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solvent specially purified and analyzed for biotechnology applications, including peptide and oligonucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deblock trichloroacetic acid soln.
Color Colorless
MDL Number MFCD00004177
UN Number UN2810
Concentration 3% w/w in dichloromethane
Density 1.325
Synonym DEBLK soln.
Quantity 2.5 L
Physical Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.