Learn More
Glen Research DEBLOCKING MIX 2000ML
Catalog No. NC9833698
Encompass_Preferred
3% Trichloroacetic acid/Dichloromethane, 2L
Catalog No. NC9833698 Supplier Glen Research Supplier No. 40414062
3% Trichloroacetic acid/Dichloromethane, 2L