Learn More
Contec™ Debris Screen for Contec Polypropylene Buckets
BIOPHARMAGREENER_CHOICE
Catalog No. 19169124 Shop All Contec Inc. Products
Catalog No. 19-169-124 Supplier Contec™ Supplier No. 2675
Add to Cart
Add to Cart
Contec Debris Screen is to be positioned in the base of a Contec polypropylene bucket to prevent the mop from touching the bottom of the bucket where unwanted contaminants settle
Specifications
Autoclavable Autoclavable