Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Debrisoquin hemisulfate | 581-88-4 | MFCD00153791 | 99.0% | 224.27 g/mol | C10H13N3.1/2H2O4S | 10 MG
SDP

Catalog No. 5000298048
Encompass_Preferred

Debrisoquin hemisulfate is the hemisulfate salt of debrisoquin (isocaramidine) supplied as a light yellow to yellow solid for research use. It has been reported to act as a TMPRSS2 inhibitor and is used in biochemical and pharmacological studies.

  • Acts as a TMPRSS2 inhibitor in cell-based entry assays
  • Light yellow to yellow solid suitable for handling and formulation
  • Molecular weight 224.27 g/mol
  • High purity (~99.0%; select batches 99.94%)
  • Storage: powder -20°C (stable 3 years); in solvent -80°C (6 months) or -20°C (1 month)
  • CAS number 581-88-4 for chemical identification

Catalog No. 50-002-98048 Supplier Medchemexpress LLC Supplier No. HYB1624A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Debrisoquin hemisulfate is the hemisulfate salt of debrisoquin (isocaramidine) supplied as a light yellow to yellow solid for research use. It has been reported to act as a TMPRSS2 inhibitor and is used in biochemical and pharmacological studies.

  • Acts as a TMPRSS2 inhibitor in cell-based entry assays
  • Light yellow to yellow solid suitable for handling and formulation
  • Molecular weight 224.27 g/mol
  • High purity (~99.0%; select batches 99.94%)
  • Storage: powder -20°C (stable 3 years); in solvent -80°C (6 months) or -20°C (1 month)
  • CAS number 581-88-4 for chemical identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.