Learn More
Medchemexpress LLC DEC-205/CD205 Human 5ug
SDP
Catalog No. 5000178670
Encompass_Preferred
DEC-205/CD205 Protein an endocytic receptor transports captured antigens to a specialized processing compartment placing it at the forefront of antigen presentation This role is crucial for immune surveillance and initiating immune responses DEC-205/CD205 s involvement also downregulates B-lymphocyte proliferation showcasing its regulatory impact on immune cell dynamics and responses DEC-205/CD205 Protein Human (HEK293 His) is the recombinant human-derived DEC-205/CD205 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78670 Supplier Medchemexpress LLC Supplier No. HYP742035UG
DEC-205/CD205 Protein an endocytic receptor transports captured antigens to a specialized processing compartment placing it at the forefront of antigen presentation This role is crucial for immune surveillance and initiating immune responses DEC-205/CD205 s involvement also downregulates B-lymphocyte proliferation showcasing its regulatory impact on immune cell dynamics and responses DEC-205/CD205 Protein Human (HEK293 His) is the recombinant human-derived DEC-205/CD205 protein expressed by HEK293 with C-His labeled tag