Learn More
Medchemexpress LLC DEC-205/CD205 Mouse 50ug | 50ug
SDP
Catalog No. 5000195328
Encompass_Preferred
DEC-205/CD205 protein acts as an endocytic receptor directing antigens to an antigen-processing compartment It also reduces B lymphocyte proliferation DEC-205/CD205 Protein Mouse (HEK293 His) is the recombinant mouse-derived DEC-205/CD205 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-95328 Supplier Medchemexpress LLC Supplier No. HYP7799250UG
DEC-205/CD205 protein acts as an endocytic receptor directing antigens to an antigen-processing compartment It also reduces B lymphocyte proliferation DEC-205/CD205 Protein Mouse (HEK293 His) is the recombinant mouse-derived DEC-205/CD205 protein expressed by HEK293 with C-His labeled tag