Learn More
Medchemexpress LLC DEC-RVRK-CMK TFA | 96.1% | 744.41 | 1 MG
SDP
Catalog No. 5000398274
Encompass_Preferred
DEC-RVRK-CMK (Decanoyl-Arg-Val-Arg-Lys-chloromethylketone) TFA is a peptide-based CMK (chloromethylketone) inhibitor that targets and inactivates the secreted soluble kexin (Kex2) (Ki=8.45 μM). The yeast enzyme Kex2 (kexin, EC 3.4.21.61) is a calcium-dependent transmembrane protease and belongs to the mammalian protease family of the serine protease subtilisin family. The binding mechanism of Kex2 with different CMK inhibitors depends on substrate selectivity, particularly the selective differences between lysine and arginine at the P1 position. It is for research use only.
  • Peptide-based CMK inhibitor
  • Targets and inactivates secreted soluble kexin (Kex2)
  • Ki of 8.45 μM against ssKex2
  • Inhibits yeast enzyme Kex2 (kexin, EC 3.4.21.61)
  • Calcium-dependent transmembrane protease
  • Useful for studying binding mechanisms and substrate selectivity
Catalog No. 50-003-98274 Supplier Medchemexpress LLC Supplier No. HYP5062A1MG
DEC-RVRK-CMK (Decanoyl-Arg-Val-Arg-Lys-chloromethylketone) TFA is a peptide-based CMK (chloromethylketone) inhibitor that targets and inactivates the secreted soluble kexin (Kex2) (Ki=8.45 μM). The yeast enzyme Kex2 (kexin, EC 3.4.21.61) is a calcium-dependent transmembrane protease and belongs to the mammalian protease family of the serine protease subtilisin family. The binding mechanism of Kex2 with different CMK inhibitors depends on substrate selectivity, particularly the selective differences between lysine and arginine at the P1 position. It is for research use only.
  • Peptide-based CMK inhibitor
  • Targets and inactivates secreted soluble kexin (Kex2)
  • Ki of 8.45 μM against ssKex2
  • Inhibits yeast enzyme Kex2 (kexin, EC 3.4.21.61)
  • Calcium-dependent transmembrane protease
  • Useful for studying binding mechanisms and substrate selectivity