Learn More
Strem, An Ascensus Company CAS# 17702-41-9. 250mg. Decaborane, min. 97%. MFCD00011314
SDP
Catalog No. 5090108693
Encompass
CAS# 17702-41-9. 250mg. Decaborane, min. 97%. MFCD00011314. Molecular Weight: 122.21. Molecular Formula: B10H14. Color/form: white xtl. Strem# 05-0500. http://www.strem.com/catalog/v/05-0500/
Catalog No. 50-901-08693 Supplier Strem, An Ascensus Company Supplier No. 050500250MG
CAS# 17702-41-9. 250mg. Decaborane, min. 97%. MFCD00011314. Molecular Weight: 122.21. Molecular Formula: B10H14. Color/form: white xtl. Strem# 05-0500. http://www.strem.com/catalog/v/05-0500/