Learn More
AccuStandard, Inc DECACHLOROBIPHENYL TCMX 1MLDE
SDP
Catalog No. NC9129900
Encompass
Decachlorobiphenyl and TCMX, 1mL, Concentration: 0.2mg/mL in Acetone, No. of Components: 2 comps., Components: Decachlorobiphenyl, CAS No.: 2051-24-3, Tetrachloro-m-xylene, CAS No.: 877-09-8
Catalog No. NC9129900 Supplier AccuStandard, Inc Supplier No. CLP032R
Decachlorobiphenyl and TCMX, 1mL, Concentration: 0.2mg/mL in Acetone, No. of Components: 2 comps., Components: Decachlorobiphenyl, CAS No.: 2051-24-3, Tetrachloro-m-xylene, CAS No.: 877-09-8