Learn More
Medchemexpress LLC Decaethylene glycol | 5579-66-8 | 99.1% | C20H42O11 | 5 G
SDP
Catalog No. 5000355823
Encompass_Preferred
Decaethylene glycol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Exploits the ubiquitin-proteasome system for targeted protein degradation.
Catalog No. 50-003-55823 Supplier Medchemexpress LLC Supplier No. HY1412325G
Decaethylene glycol is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This compound leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Exploits the ubiquitin-proteasome system for targeted protein degradation.