Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decaethylene glycol | 5579-66-8 | 99.13% | C20H42O11 | 1 G
SDP

Catalog No. 5000355822
Encompass_Preferred

Decaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 458.54
  • Appearance: <25.8°C Solid, >28.3°C Liquid
  • Color: Colorless to off-white
  • Solubility: In DMSO: 100 mg/mL (218.08 mM)
  • Target: PROTAC linkers
  • Pathway: PROTAC

Catalog No. 50-003-55822 Supplier Medchemexpress LLC Supplier No. HY1412321G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decaethylene glycol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 458.54
  • Appearance: <25.8°C Solid, >28.3°C Liquid
  • Color: Colorless to off-white
  • Solubility: In DMSO: 100 mg/mL (218.08 mM)
  • Target: PROTAC linkers
  • Pathway: PROTAC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.