Learn More
Medchemexpress LLC Decaethylene glycol dodecyl ether | 618113-48-7 | MFCD00871338 | 99.9% | 626.86 g/mol | C32H66O11 | 5 MG
SDP
Catalog No. 5000262144
Encompass_Preferred
Decaethylene glycol dodecyl ether is a PEG-based, nonionic alkyl ether used as a flexible linker or spacer in PROTAC synthesis and bioconjugation. It combines a dodecyl (C12) hydrophobic tail with a decaethylene glycol spacer to balance solubility and hydrophobicity for bifunctional molecules.
  • Useful as a PEG-based PROTAC linker and spacer.
  • Provides a hydrophobic C12 tail with a decaethylene glycol spacer.
  • High purity (≈99.9%) suitable for synthetic applications.
  • Molecular weight approximately 626.86 g/mol for planning stoichiometry.
  • Available in small research quantities for synthesis trials.
Catalog No. 50-002-62144 Supplier Medchemexpress LLC Supplier No. HY1320655MG
Decaethylene glycol dodecyl ether is a PEG-based, nonionic alkyl ether used as a flexible linker or spacer in PROTAC synthesis and bioconjugation. It combines a dodecyl (C12) hydrophobic tail with a decaethylene glycol spacer to balance solubility and hydrophobicity for bifunctional molecules.
  • Useful as a PEG-based PROTAC linker and spacer.
  • Provides a hydrophobic C12 tail with a decaethylene glycol spacer.
  • High purity (≈99.9%) suitable for synthetic applications.
  • Molecular weight approximately 626.86 g/mol for planning stoichiometry.
  • Available in small research quantities for synthesis trials.