Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decaethylene glycol dodecyl ether | 618113-48-7 | MFCD00871338 | 99.9% | C32H66O11 | 10MG
SDP

Catalog No. 5000260691
Encompass_Preferred

Decaethylene glycol dodecyl ether is a PEG-based linker (PEG10) featuring a dodecyl alkyl tail and a decaethylene glycol chain, used as a spacer in PROTAC synthesis and conjugation chemistries. It is supplied as a high-purity material with specified storage conditions to preserve stability.

  • High purity (≈99.9%).
  • Molecular weight 626.86 g/mol.
  • Chemical formula C32H66O11.
  • Appearance: white to off-white oil.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-60691 Supplier Medchemexpress LLC Supplier No. HY13206510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decaethylene glycol dodecyl ether is a PEG-based linker (PEG10) featuring a dodecyl alkyl tail and a decaethylene glycol chain, used as a spacer in PROTAC synthesis and conjugation chemistries. It is supplied as a high-purity material with specified storage conditions to preserve stability.

  • High purity (≈99.9%).
  • Molecular weight 626.86 g/mol.
  • Chemical formula C32H66O11.
  • Appearance: white to off-white oil.
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.