Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobenzophenone, 97%

Catalog No. AAA1926206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1926206 5 g
AAA1926203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1926206 Supplier Thermo Scientific Chemicals Supplier No. A1926206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 853-39-4 | C13F10O | 362.126 g/mol

Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853-39-4
Molecular Formula C13F10O
Molecular Weight (g/mol) 362.126
MDL Number MFCD00000295
InChI Key WWQLXRAKBJVNCC-UHFFFAOYSA-N
Synonym decafluorobenzophenone, benzophenone, decafluoro, perfluorobenzophenone, methanone, bis pentafluorophenyl, bis perfluorophenyl methanone, bis pentafluorophenyl methanone, bis 2,3,4,5,6-pentafluorophenyl methanone, di2,3,4,5,6-pentafluorophenyl ketone, acmc-209q5n
PubChem CID 70068
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)methanone
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C(=O)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
Beilstein 2066476
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 362.12
Percent Purity 98%
Chemical Name or Material Decafluorobenzophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-717-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.