Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobiphenyl, 99%

Catalog No. AAA1594806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594806 5 g
AAA1594814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1594806 Supplier Thermo Scientific Chemicals Supplier No. A1594806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 434-90-2 | C12F10 | 334.116 g/mol

Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decafluorobiphenyl is used as an intermediate in organic synthesis and in pharmaceuticals.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-90-2
Molecular Formula C12F10
Molecular Weight (g/mol) 334.116
MDL Number MFCD00000292
InChI Key ONUFSRWQCKNVSL-UHFFFAOYSA-N
Synonym decafluorobiphenyl, perfluorobiphenyl, biphenyl, decafluoro, perfluorodiphenyl, 1,1'-biphenyl, 2,2',3,3',4,4',5,5',6,6'-decafluoro, 1,2,3,4,5-pentafluoro-6-2,3,4,5,6-pentafluorophenyl benzene, decafluoro biphenyl, pubchem9114, acmc-1ctuw
PubChem CID 67949
IUPAC Name 1,2,3,4,5-pentafluoro-6-(2,3,4,5,6-pentafluorophenyl)benzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)C2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 66°C to 70°C
Density 1.785
Boiling Point 206°C
Quantity 5 g
Beilstein 1916978
Solubility Information Insoluble in water.
Formula Weight 334.12
Percent Purity 99%
Chemical Name or Material Decafluorobiphenyl
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
DOTInformation Hazard Class: 9; Packaging Group: II
EINECSNumber 207-107-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.