Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorotriphenylphosphine, MilliporeSigma™ Supelco™

Catalog No. 111014378 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4378 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4378 Supplier MilliporeSigma Supelco Supplier No. 442543
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decafluorotriphenylphosphine (DFTPP) is commonly used as a check standard for the calibration of mass spectrum detector (MSD) prior to the analysis of samples under investigation.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 5074-71-5
Flash Point 113°C
Physical Form Neat
Linear Formula C6H5 P(C6F5)2
MDL Number MFCD00000291
Melting Point 55°C to 60°C
Synonym Bis(pentafluorophenyl)phenylphosphine; DFTPP; 2,3,4,5,6,2 ′,3 ′,4 ′,5 ′,6 ′-Decafluorotriphenylphosphine
Type Food and Cosmetic Component Standard
Quantity 100 mg
Formula Weight 442.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.