Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorotriphenylphosphine Solution, 250μg/mL in dichloromethane, Ultra Scientific

Catalog No. USIST3401
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-3401 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-3401 Supplier Agilent Technologies Supplier No. IST3401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5074-71-5 , 75-09-2
Molecular Formula C18H5F10P
Molecular Weight (g/mol) 442.20
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
Synonym DFTPP
PubChem CID 78757
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)(phenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

CAS Max % 99.98, 0.02
Physical Form Liquid
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 250μg/mL
Chemical Name or Material Decafluorotriphenylphosphine Solution (DFTPP), 250μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.