Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decahydroisoquinoline (cis- and trans- mixture, predominantly cis-isomer) 98.0+%, TCI America™
SDP

Catalog No. D50675ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50675ML 5 mL
D506725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50675ML Supplier TCI America Supplier No. D50675ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6329-61-9
Molecular Formula C9H17N
Molecular Weight (g/mol) 139.24
MDL Number MFCD00012096
InChI Key NENLYAQPNATJSU-UHFFFAOYNA-N
PubChem CID 97812
IUPAC Name decahydroisoquinoline
SMILES C1CCC2CNCCC2C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 mL
Formula Weight 139.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Decahydroisoquinoline (cis- and trans- mixture, predominantly cis-isomer)
EINECSNumber (5)-6524
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.