Learn More
StatLab™ Decal Decalcifier
GSA_VASDP
Catalog No. STL121132
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 32 oz. Bottle
5 gal. Cube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL121132 4 x 32 oz. Bottle
STL12111 4 x 1 gal.
STL12115 5 gal. Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL121132 Supplier StatLab™ Supplier No. 121132
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water.
  • Can be used to safely decalcify all types of tissue samples, from needle biopsies to cortical bone
  • Mild enough that most samples can be left in overnight
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 32 oz. Bottle