Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Decalactone | 706-14-9 | 100 G
SDP

Catalog No. 5000417647
Encompass_Preferred

γ-Decalactone is a natural flavoring substance with a fruity peach aroma, commonly used as a food additive. This colorless to light yellow liquid has a density of 0.948 g/cm³, a molecular weight of 170.25, and a chemical formula of C10H18O2. It is initially sourced from tropical yeast strains like Yarrowia lipolytica CCMA 0242 and Lindnera saturnus CCMA 0243, with Ricinoleic acid often used as a substrate in its production.

  • Natural flavoring substance
  • Fruity aroma of juicy peaches
  • Can be used as a food additive
  • Colorless to light yellow liquid appearance
  • Density of 0.948 g/cm³
  • Molecular weight of 170.25
  • Chemical formula C10H18O2
  • Derived from microorganisms
  • For research use only

Catalog No. 50-004-17647 Supplier Medchemexpress LLC Supplier No. HYN7105100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Decalactone is a natural flavoring substance with a fruity peach aroma, commonly used as a food additive. This colorless to light yellow liquid has a density of 0.948 g/cm³, a molecular weight of 170.25, and a chemical formula of C10H18O2. It is initially sourced from tropical yeast strains like Yarrowia lipolytica CCMA 0242 and Lindnera saturnus CCMA 0243, with Ricinoleic acid often used as a substrate in its production.

  • Natural flavoring substance
  • Fruity aroma of juicy peaches
  • Can be used as a food additive
  • Colorless to light yellow liquid appearance
  • Density of 0.948 g/cm³
  • Molecular weight of 170.25
  • Chemical formula C10H18O2
  • Derived from microorganisms
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.