Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Decalactone | 706-14-9 | 50 G
SDP

Catalog No. 5000418102
Encompass_Preferred

γ-Decalactone is a natural flavoring substance characterized by its fruity peach aroma. It is commonly used as a food additive and for general laboratory research. This compound is also utilized in the manufacturing of various substances, with Ricinoleic acid often employed as a substrate in its production.

  • Natural flavoring substance
  • Fruity peach aroma
  • Appearance: colorless to light yellow liquid
  • High purity: 99.36%
  • Molecular formula: C10H18O2
  • Molecular weight: 170.25
  • Stable under recommended storage conditions
  • Identified for use in laboratory chemicals
  • Used in the manufacture of substances

Catalog No. 50-004-18102 Supplier Medchemexpress LLC Supplier No. HYN710550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

γ-Decalactone is a natural flavoring substance characterized by its fruity peach aroma. It is commonly used as a food additive and for general laboratory research. This compound is also utilized in the manufacturing of various substances, with Ricinoleic acid often employed as a substrate in its production.

  • Natural flavoring substance
  • Fruity peach aroma
  • Appearance: colorless to light yellow liquid
  • High purity: 99.36%
  • Molecular formula: C10H18O2
  • Molecular weight: 170.25
  • Stable under recommended storage conditions
  • Identified for use in laboratory chemicals
  • Used in the manufacture of substances

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.