Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC γ-Decalactone | 706-14-9 | 500 G
SDP

Supplier:  Medchemexpress LLC HYN7105500G

Encompass_Preferred

γ-Decalactone is a natural flavoring substance with the fruity aroma of juicy peaches. It can be used as a food additive and is identified as a laboratory chemical used in the manufacture of substances.

  • Natural flavoring substance
  • Fruity aroma of juicy peaches
  • Can be used as a food additive
  • Purity: 99.36%
  • Molecular weight: 170.25
  • Chemical formula: C10H18O2
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Stable under recommended storage conditions
  • For research use only

Catalog No. 50-004-19999


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.