Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Decalactone, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015302 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5302 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5302 Supplier MilliporeSigma Supelco Supplier No. 937381ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

γ-Decalactone is a flavor and fragrance compound, which finds applications in food and beverage industry.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 706-14-9
Density 0.948 g/mL (at 25°C)
Flash Point 136°C
Physical Form Neat
Linear Formula C10H18O2
MDL Number MFCD00005404
Synonym gamma-Decanolactone; 4-Hydroxydecanoic acid gamma-lactone
Type Analytical Standard
Quantity 1 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.