Learn More
Electron Microscopy Sciences Hard Specimen Decalcifier 1 LT
SDP
Catalog No. 5025292
Encompass_Preferred
Ready-to-use decalcifier is ideal for hard specimens and protects the cellular structure with minial interference with subsequent staining. For use with human and animal calcified tissue, bone, cartilage and other hard specimens.
Catalog No. 50-252-92 Supplier Electron Microscopy Sciences Supplier No. 6414706
Ready-to-use decalcifier is ideal for hard specimens and protects the cellular structure with minial interference with subsequent staining. For use with human and animal calcified tissue, bone, cartilage and other hard specimens.