Learn More
Ethos Biosciences Decalcifier Formic Acid 1gal
Catalog No. 5000819170
Encompass_Preferred
Formic Acid Decalcifier is a clear colorless to pale yellow aqueous solution formulated for histological decalcification of bone and calcified tissues Contains 10% formic acid in a stable ready to use formulation that gentley removes calcium salts while preserving tissue morpholoy
Catalog No. 50-008-19170 Supplier Ethos Biosciences Supplier No. 3360G
Formic Acid Decalcifier is a clear colorless to pale yellow aqueous solution formulated for histological decalcification of bone and calcified tissues Contains 10% formic acid in a stable ready to use formulation that gentley removes calcium salts while preserving tissue morpholoy