Learn More
Leica Biosystems DECALCIFIER I 1 GALLON
Catalog No. NC9903324
Encompass_Preferred
Decalcifier I is a unique decalcifying solution that has the ability to fix and decalcify at the same time.
Catalog No. NC9903324 Supplier Leica Biosystems Supplier No. 3800440
Decalcifier I is a unique decalcifying solution that has the ability to fix and decalcify at the same time.