Learn More
Leica Biosystems DECALCIFIER II GALLON
Supplier:  Leica Biosystems 3800460
Encompass_Preferred
Decalcifier I is a unique decalcifying solution that has the ability to fix and decalcify at the same time.
Catalog No. 50-255-2444