Learn More
Ethos Biosciences Disodium EDTA 10%, 4oz, Ethos
Catalog No. 5000574217
Encompass_Preferred
Disodium EDTA 10% is a ready-to-use chelating solution. Commonly used for gentle decalcification of bone and calcified tissue specimens, it helps preserve cellular morphology and antigenicity for reliable microscopic evaluation and special staining procedures. The solution provides consistent, controlled decalcification, making it ideal for surgical pathology, biopsy, and research samples. It is also suitable for metal ion chelation, buffer preparation, and other routine laboratory tasks. Supplied in chemically compatible containers, Disodium EDTA 10% is appropriate for clinical, research, pharmaceutical, and educational laboratories.
Catalog No. 50-005-74217 Supplier Ethos Biosciences Supplier No. 336604
Disodium EDTA 10% is a ready-to-use chelating solution. Commonly used for gentle decalcification of bone and calcified tissue specimens, it helps preserve cellular morphology and antigenicity for reliable microscopic evaluation and special staining procedures. The solution provides consistent, controlled decalcification, making it ideal for surgical pathology, biopsy, and research samples. It is also suitable for metal ion chelation, buffer preparation, and other routine laboratory tasks. Supplied in chemically compatible containers, Disodium EDTA 10% is appropriate for clinical, research, pharmaceutical, and educational laboratories.