Learn More
StatLab Decalcifying Fixative - HCl-Formic Acid in Formalin
Description
- Used to both fix (preserve) and decalcify (demineralize) hard mineralized tissues such as bone, teeth, or calcified biopsies
- Solution dissolves hard calcium and phosphorus mineral deposits (hydroxyapatite) from bone and teeth
Specifications
Specifications
| Product Type | Decalcifying/Fixation Solution |
| Quantity | 1 gal. |
| Solution Type | Decalcifier |
| Form | Solution |