Learn More
Newcomer Supply Decalcifying Solution, HCL/EDTA, 1 gallon
SDP
Catalog No. NC1606263
Encompass
Use: Excellent for bone marrows. Gentle but quick! Newcomer Supply Decalcifying Solution, HCL/ EDTA, combines acid and chelating decalcifying agents, with an added tartrate buffer for prevention of cellular swelling and distortion. This solution provides rapid decalcification, maintains excellent cellular morphology and is suitable for all bone specimens from bone marrow biopsies and disc material (light bone) to femoral head and long bone sections (compact bone). It is not recommended for use when proteoglycan preservation in articular cartilage is important. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC1606263 Supplier Newcomer Supply Supplier No. 1050C
Use: Excellent for bone marrows. Gentle but quick! Newcomer Supply Decalcifying Solution, HCL/ EDTA, combines acid and chelating decalcifying agents, with an added tartrate buffer for prevention of cellular swelling and distortion. This solution provides rapid decalcification, maintains excellent cellular morphology and is suitable for all bone specimens from bone marrow biopsies and disc material (light bone) to femoral head and long bone sections (compact bone). It is not recommended for use when proteoglycan preservation in articular cartilage is important. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.