Learn More
Newcomer Supply Decalcifying Solution, EDTA/Sucrose, 10 Liter Cube
SDP
Catalog No. NC1197229
Encompass
Newcomer Supply Decalcifying Solution, EDTA/Sucrose procedure uses a chelating agent with an added TRIS buffer for gentle bone decalcification.  Decalcification rate will be slower but preservation of cellular morphology is excellent and viability of staining for enzymes, immunohistochemistry antigenicity and electron microscopy is maintained. This solution is not recommended for use when proteoglycan preservation in articular cartilage is important. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1197229 Supplier Newcomer Supply Supplier No. 1048D
Newcomer Supply Decalcifying Solution, EDTA/Sucrose procedure uses a chelating agent with an added TRIS buffer for gentle bone decalcification.  Decalcification rate will be slower but preservation of cellular morphology is excellent and viability of staining for enzymes, immunohistochemistry antigenicity and electron microscopy is maintained. This solution is not recommended for use when proteoglycan preservation in articular cartilage is important. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.