Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decalcifying Solution, Krajian, for Decalcification of Bones, Ricca Chemical
SDP

Catalog No. 254516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2545-16 500 mL Natural Poly Bottle
2545-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2545-16 Supplier Ricca Chemical Company Supplier No. 254516
Add to Cart
Add to Cart

For Decalcification of Bones

Specifications

Physical Form Liquid
Chemical Name or Material Decalcifying Solution
Name Note Krajian
CAS 7732-18-5, 64-18-6, 6132-04-3
CAS Min % 70.0, 21.27, 6.98
CAS Max % 73.0, 22.14, 7.26
Packaging Natural Poly Bottle
Color Colorless
Quantity 500 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.