Learn More
Decalcifying Solution, Krajian, for Decalcification of Bones, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Physical Form | Liquid |
| Chemical Name or Material | DECALCIFYING SOLUTION |
| CAS | 7732-18-5, 64-18-6, 6132-04-3 |
| CAS Min % | 70.0, 21.27, 6.98 |
| CAS Max % | 73.0, 22.14, 7.26 |
| Packaging | Natural Poly Bottle |
| Color | Colorless |
| Quantity | 1 L |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.