Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cambridge Isotope Laboratories Decalin (cis/trans mix) (D18 99%) 5 g

Supplier:  Cambridge Isotope Laboratories DLM13865

Encompass

Decalin (cis/trans mix) (D18 99%) 5 g

Catalog No. 50-001-68529


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.