Learn More
Apexbio Technology LLC Decamethonium Bromide 541-22-0 5g
SDP
Catalog No. 5000092642
Encompass_Preferred
Decamethonium Bromide (CAS 541-22-0) is a bis-quaternary ammonium compound targeting nicotinic acetylcholine receptors (nAChRs) at the neuromuscular junction It is designed to bind and activate muscle-type nAChRs thereby inducing sustained depolarization and neuromuscular blockade Decamethonium Bromide exerts its biological activity primarily through partial agonist-mediated depolarization block of these receptors preventing normal muscle contractions Based on these pharmacological properties Decamethonium Bromide holds research potential in the evaluation of neuromuscular blockers and in studies elucidating receptor-channel function in neuromuscular transmission
Catalog No. 50-000-92642 Supplier Apexbio Technology LLC Supplier No. B16015000
Decamethonium Bromide (CAS 541-22-0) is a bis-quaternary ammonium compound targeting nicotinic acetylcholine receptors (nAChRs) at the neuromuscular junction It is designed to bind and activate muscle-type nAChRs thereby inducing sustained depolarization and neuromuscular blockade Decamethonium Bromide exerts its biological activity primarily through partial agonist-mediated depolarization block of these receptors preventing normal muscle contractions Based on these pharmacological properties Decamethonium Bromide holds research potential in the evaluation of neuromuscular blockers and in studies elucidating receptor-channel function in neuromuscular transmission