Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decamethonium Bromide | 541-22-0 | 98.7% | 5 G
SDP

Catalog No. 5000408668
Encompass_Preferred

Decamethonium Bromide is an acetylcholine receptor inhibitor and muscle relaxant. It functions by first inducing depolarization of skeletal muscles, followed by binding to postsynaptic acetylcholine receptors to cause persistent paralysis. This product is intended for research use only and is not sold to patients.

  • Inhibits acetylcholine receptors and acts as a muscle relaxant
  • Initiates muscle depolarization before inducing paralysis
  • Suitable for various research applications in pharmacology and neuroscience
  • Soluble in DMSO (83.33 mg/mL) and H2O (≥ 50 mg/mL)
  • Recommended storage at 4°C sealed, away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-08668 Supplier Medchemexpress LLC Supplier No. HYB0570A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decamethonium Bromide is an acetylcholine receptor inhibitor and muscle relaxant. It functions by first inducing depolarization of skeletal muscles, followed by binding to postsynaptic acetylcholine receptors to cause persistent paralysis. This product is intended for research use only and is not sold to patients.

  • Inhibits acetylcholine receptors and acts as a muscle relaxant
  • Initiates muscle depolarization before inducing paralysis
  • Suitable for various research applications in pharmacology and neuroscience
  • Soluble in DMSO (83.33 mg/mL) and H2O (≥ 50 mg/mL)
  • Recommended storage at 4°C sealed, away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.