Learn More
Medchemexpress LLC Decamethonium (Bromide) | 541-22-0 | 99.6% | 1 G
SDP
Catalog No. 5000408379
Encompass_Preferred
Decamethonium Bromide is a biochemical reagent used in life science research. It acts as an acetylcholine receptor inhibitor and muscle relaxant, initially inducing skeletal muscle depolarization before binding to postsynaptic acetylcholine receptors to cause persistent paralysis.
  • Biochemical reagent for life science research.
  • Functions as an acetylcholine receptor inhibitor.
  • Acts as a muscle relaxant.
  • Has a purity of 99.6%.
  • Targets nAChR.
  • Involved in membrane transporter/ion channel and neuronal signaling pathways.
Catalog No. 50-004-08379 Supplier Medchemexpress LLC Supplier No. HYB0570A1G
Decamethonium Bromide is a biochemical reagent used in life science research. It acts as an acetylcholine receptor inhibitor and muscle relaxant, initially inducing skeletal muscle depolarization before binding to postsynaptic acetylcholine receptors to cause persistent paralysis.
  • Biochemical reagent for life science research.
  • Functions as an acetylcholine receptor inhibitor.
  • Acts as a muscle relaxant.
  • Has a purity of 99.6%.
  • Targets nAChR.
  • Involved in membrane transporter/ion channel and neuronal signaling pathways.