Learn More
Medchemexpress LLC Decamethonium Bromide | 541-22-0 | MFCD00011779 | 98.7% | 1 ML
SDP
Catalog No. 5000407863
Encompass_Preferred
Decamethonium Bromide is an acetylcholine receptor inhibitor and muscle relaxant. It functions by inducing depolarization of skeletal muscles and subsequently binding to postsynaptic acetylcholine receptors, leading to persistent paralysis. For research use only.
  • Acetylcholine receptor inhibitor
  • Muscle relaxant
  • Induces depolarization of skeletal muscles
  • Binds to postsynaptic acetylcholine receptors to induce persistent paralysis
Catalog No. 50-004-07863 Supplier Medchemexpress LLC Supplier No. HYB057010MM/1ML
Decamethonium Bromide is an acetylcholine receptor inhibitor and muscle relaxant. It functions by inducing depolarization of skeletal muscles and subsequently binding to postsynaptic acetylcholine receptors, leading to persistent paralysis. For research use only.
  • Acetylcholine receptor inhibitor
  • Muscle relaxant
  • Induces depolarization of skeletal muscles
  • Binds to postsynaptic acetylcholine receptors to induce persistent paralysis