Learn More
Selleck Chemical LLC Decamethonium Bromide S4072-50mg
SDP
Catalog No. 503108697
Encompass_Preferred
Decamethonium Bromide is a nicotinic AChR partial agonist and neuromuscular blocking agent
Catalog No. 50-310-8697 Supplier Selleck Chemical LLC Supplier No. S407250MG
Decamethonium Bromide is a nicotinic AChR partial agonist and neuromuscular blocking agent