Learn More
Medchemexpress LLC Decamethoxine 250mg | 38146-42-8 | 250MG
SDP
Catalog No. 5000241834
Encompass_Preferred
Decamethoxine (Septefril) is a cationic gemini surfactant Decamethoxine exhibits strong bactericidal and fungicidal effects Decamethoxine modifies the permeability of the microbial cell membrane resulting in the destruction and death of diverse microorganisms[1 [2
Catalog No. 50-002-41834 Supplier Medchemexpress LLC Supplier No. HY108004250MG
Add to Cart
Add to Cart
Decamethoxine (Septefril) is a cationic gemini surfactant Decamethoxine exhibits strong bactericidal and fungicidal effects Decamethoxine modifies the permeability of the microbial cell membrane resulting in the destruction and death of diverse microorganisms[1 [2