Learn More
Medchemexpress LLC Decamethoxine | 38146-42-8 | MFCD02654058 | 95.0% | 693.91 g/mol | C38H74Cl2N2O4 | 10 G
SDP
Catalog No. 5000242859
Encompass_Preferred
Decamethoxine (Septefril) is a cationic gemini surfactant with broad-spectrum bactericidal and fungicidal activity. It disrupts microbial cell membrane permeability leading to rapid death of susceptible microorganisms, and has been reported to reduce bacterial adhesion at concentrations around 10 μg/mL. Supplied as a solid with 95.0% listed purity in a 10 G pack for research and formulation testing.
  • Broad-spectrum bactericidal and fungicidal activity
  • Disrupts microbial membrane permeability
  • Reduces bacterial adhesion at low concentrations
  • Supplied as a solid with high listed purity
  • Suitable for research and formulation testing
Catalog No. 50-002-42859 Supplier Medchemexpress LLC Supplier No. HY10800410G
Add to Cart
Add to Cart
Decamethoxine (Septefril) is a cationic gemini surfactant with broad-spectrum bactericidal and fungicidal activity. It disrupts microbial cell membrane permeability leading to rapid death of susceptible microorganisms, and has been reported to reduce bacterial adhesion at concentrations around 10 μg/mL. Supplied as a solid with 95.0% listed purity in a 10 G pack for research and formulation testing.
  • Broad-spectrum bactericidal and fungicidal activity
  • Disrupts microbial membrane permeability
  • Reduces bacterial adhesion at low concentrations
  • Supplied as a solid with high listed purity
  • Suitable for research and formulation testing