Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Decamethylcyclopentasiloxane

Catalog No. 444278250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-427-8250ML 250 mL
44-427-850ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-427-8250ML Supplier Sigma Aldrich Supplier No. 444278250ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Decamethylcyclopentasiloxane (D5) is used in personal care products including skin creams, cosmetics, shampoos, deodorants and conditioners. D5 is also used in various applications such as industrial cleaning fluids and dry cleaning solvents.Decamethylcyclopentasiloxane (D5) can be used as:A greener solvent in synthetic chemistry applications.A monomeric unit for polymerization by various base catalysts to obtain polysiloxane polymer.

Specifications

CAS 541-02-6
Density 0.958 g/mL at 25°C
Boiling Point 90°C (10 mmHg, lit.)
Molecular Formula C10H30O5Si5
Refractive Index n20/D 1.396
Linear Formula C10H30O5Si5
MDL Number MFCD00046966
Quantity 250 mL
Molecular Weight (g/mol) 370.77 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.