Learn More
Medchemexpress LLC Decamethylenebispyridinium dibromide | 6266-40-6 | MFCD00031922 | 99.7% | 458.27 g·mol⁻¹ | C20H30Br2N2 | 100 MG
SDP
Catalog No. 5000280558
Encompass_Preferred
1,10-Bis(pyridinium)decane (Decamethylenebispyridinium dibromide) is a biscationic small molecule AChE inhibitor supplied as a solid for biochemical and pharmacological research. It is used primarily in vitro to probe acetylcholinesterase activity and cholinergic pathways.
  • Binds and inhibits acetylcholinesterase (AChE) with an IC50 of 25.8 nM.
  • High purity (99.66%) suitable for research applications.
  • Chemical formula C20H30Br2N2 and molecular weight 458.27 g·mol⁻¹.
  • White to off-white solid, convenient handling and storage.
  • Available in multiple pack sizes, including 100 MG.
  • Intended for biochemical assays to study cholinesterase-related mechanisms.
Catalog No. 50-002-80558 Supplier Medchemexpress LLC Supplier No. HYW165085100MG
1,10-Bis(pyridinium)decane (Decamethylenebispyridinium dibromide) is a biscationic small molecule AChE inhibitor supplied as a solid for biochemical and pharmacological research. It is used primarily in vitro to probe acetylcholinesterase activity and cholinergic pathways.
  • Binds and inhibits acetylcholinesterase (AChE) with an IC50 of 25.8 nM.
  • High purity (99.66%) suitable for research applications.
  • Chemical formula C20H30Br2N2 and molecular weight 458.27 g·mol⁻¹.
  • White to off-white solid, convenient handling and storage.
  • Available in multiple pack sizes, including 100 MG.
  • Intended for biochemical assays to study cholinesterase-related mechanisms.