Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanal, 96%

Catalog No. p-7081613
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11656AE 100 mL
AAA11656AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11656AE Supplier Thermo Scientific Chemicals Supplier No. A11656AE
Only null left
Add to Cart

CAS: 112-31-2 | C10H20O | 156.269 g/mol

Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decanal is used in fragrances and flavoring and it is an important component in citrus along with octanal, citral, and sinensal. It is also used in Air Care Products, Laundry and Dishwashing Products, Personal Care Products.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-31-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00007031
InChI Key KSMVZQYAVGTKIV-UHFFFAOYSA-N
Synonym decyl aldehyde, capraldehyde, caprinaldehyde, decanaldehyde, n-decyl aldehyde, capric aldehyde, 1-decanal, n-decanal, n-decaldehyde, 1-decyl aldehyde
PubChem CID 8175
ChEBI CHEBI:31457
IUPAC Name decanal
SMILES CCCCCCCCCC=O

Specifications

Melting Point °C to 5°C
Density 0.828
Boiling Point 207°C to 209°C
Flash Point 85°C (185°F)
Assay Percent Range 96%
Refractive Index 1.428
Linear Formula CH3(CH2)8CHO
Quantity 100 mL
Beilstein 1362530
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 156.27
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Decanal
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P501c
EINECSNumber 203-957-4
RTECSNumber HD6000000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.