Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanamide, N-[(4-hydroxy-3-methoxyphenyl)methyl]- | 31078-36-1 | 5 MG
SDP

Catalog No. 5000419845
Encompass_Preferred

N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum. It significantly reduced the radical length of Lactuca sativa seedling in a dose-dependent manner. It has a molecular weight of 307.43 and a chemical formula of C18H29NO3. Its appearance is a solid, white to off-white in color. It is for research use only.

  • Capsaicinoid isolated from Capsicum annuum
  • Significantly reduced radical length of Lactuca sativa seedling
  • Molecular weight of 307.43
  • Chemical formula of C18H29NO3
  • Appears as a white to off-white solid
  • For research use only

Catalog No. 50-004-19845 Supplier Medchemexpress LLC Supplier No. HYN50995MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Vanillyldecanamide is a capsaicinoid isolated from the fruits of Capsicum annuum. It significantly reduced the radical length of Lactuca sativa seedling in a dose-dependent manner. It has a molecular weight of 307.43 and a chemical formula of C18H29NO3. Its appearance is a solid, white to off-white in color. It is for research use only.

  • Capsaicinoid isolated from Capsicum annuum
  • Significantly reduced radical length of Lactuca sativa seedling
  • Molecular weight of 307.43
  • Chemical formula of C18H29NO3
  • Appears as a white to off-white solid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.