Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Decane | 2L | 124-18-5 | MFCD00008954

Supplier:  Sigma Organic Chemistry 305702.5L

Encompass_Preferred

CAS #: 124-18-5
MDL #: MFCD00008954
Purity: >95 %
UNSPSC Code: 12352100
Molecular Weight: 142.28

Catalog No. 30-570-25L


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.