Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019651 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9651 5 mL
11-101-8370 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9651 Supplier MilliporeSigma Supelco Supplier No. 305405ML
Add to Cart
Add to Cart

Decane is an alkane hydrocarbon. It exists as a colourless flammable liquid. It is mostly used as solvent. Its vapors may be narcotic at high concentrations.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 410°F
CAS 124-18-5
Vapor Pressure 3.77 mmHg (37.7°C)
Vapor Density 4.9 (vs. air)
Density 0.73 g/mL (at 25°C)
Flash Point 57°C
Physical Form Neat
Linear Formula CH3(CH2)8 CH3
MDL Number MFCD00008954
Melting Point -30°C
Boiling Point 174°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 142.28
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.