Learn More
Supelco Inc Decane, 124-18-5, MFCD00008954, 1L
Supplier:  Supelco Inc 4571161L
Encompass
Anhydrous, ≥99%, Linear Formula: CH3(CH2)8CH3, Molecular Weight: 142.28, BP: 174 °C(lit.), MP: −30 °C (lit.). Decane has been used as an organic solvent for nanocrystal suspension. Decane-1,2-diol derivative might prevent cancer development. Decane (C10 H12) is an acyclic saturated hydrocarbon.
Catalog No. NC0569686