Learn More
Sigma Aldrich Fine Chemicals Biosciences Decane, 124-18-5, MFCD00008954, 2 L
Catalog No. NC0918593
Encompass
Linear Formula: CH3(CH2)8CH3, Molecular Weight: 142.28, anhydrous, ≥99%.Decane has been used as an organic solvent for nanocrystal suspension.
Catalog No. NC0918593 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4571162L
Linear Formula: CH3(CH2)8CH3, Molecular Weight: 142.28, anhydrous, ≥99%.Decane has been used as an organic solvent for nanocrystal suspension.